Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Potassium nitrate, 99+%, for biochemistry

Catalog No. AC447820025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447820025 2.5 kg Plastic bottle
AC447825000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447820025 Supplier Thermo Scientific Chemicals Supplier No. 447820025
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium;nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Boiling Point 400.0°C
Melting Point 334.0°C
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula KNO3
Merck Index 15,7769
Solubility Information (1 M in water) Clear colorless
Formula Weight 101.1
Percent Purity ≥99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Potassium nitrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 231-818-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.