Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium nitrate, Puriss., meets analytical spec. of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%, Solstice

Catalog No. 6001286 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-012-86 1 kg
60-012-87 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-012-86 Supplier Solstice Supplier No. 126481KG

Puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium;nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Quantity 1 kg
UN Number 1486
Formula Weight 101.1g/mol
Percent Purity 99 to 100.5%
Grade puriss.
Chemical Name or Material Potassium Nitrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
Hazard Statement P210-P220-P221-P370 + P378
Precautionary Statement H272-H412
EINECSNumber 231-818-8
RTECSNumber TT3700000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.