Learn More
Potassium osmium(VI) oxide dihydrate, 98+%
Description
Potassium osmium(VI) oxide dihydrate acts as a catalyst for the oxidative cleavage of olefins to ketones and carboxylic acids. It is employed in an intramolecular amidohydroxylation of carbamoyloxytethered olefins providing hydroxy oxazolidinones. It is also used as a reagent in the synthesis of amphidinolide B, which shows potent antitumor activity against human solid and blood tumor cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 1 g |
| Assay Percent Range | 99+% |
| Solubility Information | Soluble in water. Slowly decomposes in aqueous solution forming the tetroxide. Insoluble in alcohol,etherSoluble in water. Insoluble in alcohol and ether. |
| Formula Weight | 368.43 (332.40 Anhydrous) |
| Sensitivity | Hygroscopic |
| Appearance | Purple powder |
| Chemical Name or Material | Potassium osmium(VI) oxide dihydrate |
Safety and Handling
| Hazard Category | H272-H290-H312+H332-H314-H335 |
| Precautionary Statement | P210-P220-P221-P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c |
| DOTInformation | DOT Class: 5.1; Packaging Group: II |
| EINECSNumber | 243-247-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.