Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium p-Toluenethiosulfonate 98.0+%, TCI America™
SDP

Catalog No. T118525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T118525G 25 g
T11855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T118525G Supplier TCI America Supplier No. T118525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28519-50-8
Molecular Formula C7H7KO2S2
Molecular Weight (g/mol) 226.349
MDL Number MFCD00003547
InChI Key RUDNWZFWWJFUSF-UHFFFAOYSA-M
Synonym p-Toluenethiosulfonic Acid Potassium Salt
PubChem CID 23662126
IUPAC Name potassium;(4-methylphenyl)-oxido-oxo-sulfanylidene-$l^{6}-sulfane
SMILES CC1=CC=C(C=C1)S(=O)(=S)[O-].[K+]

Specifications

Melting Point 229°C
Color White
Quantity 25 g
Formula Weight 226.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Potassium p-Toluenethiosulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.