Learn More
Potassium perchlorate, 99%
Description
Potassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.
Solubility
Soluble in water. Slightly soluble in acetone. Insoluble in ethanol, ethyl acetate and ether.
Notes
Hygroscopic. Incompatible with strong reducing agents, powdered metals, strong acids and alcohols.

Specifications
Specifications
Melting Point | 301°C to 304°C |
Quantity | 2500 g |
UN Number | UN1489 |
Merck Index | 14,7653 |
Solubility Information | Soluble in water. Slightly soluble in acetone. Insoluble in ethanol,ethyl acetate and ether. |
IUPAC Name | potassium perchlorate |
Formula Weight | 138.55 |
Percent Purity | 99% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H271-H302 |
Hazard Statement | GHS H Statement H271-H302 May cause fire or explosion strong oxidizer. Harmful if swallowed. |
Precautionary Statement | P210-P220-P221-P264b-P270-P280-P283-P301+P312-P306+P360-P330-P370+P378q-P371+P380+P375-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: POTASSIUM PERCHLORATE |
EINECSNumber | 231-912-9 |
RTECSNumber | SC9700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.