Learn More
Potassium Periodate, ACS Reagent (Potassium Metaperiodate), Reagents
Specifications
Specifications
| Boiling Point | 300°C (loses oxygen) |
| Color | White |
| Physical Form | Solid |
| Quantity | 25 kg |
| Grade | ACS |
| Packaging | Fiber Drum |
| Specific Gravity | 3.618 |
| Chemical Name or Material | Potassium Periodate, ACS Reagent (Potassium Metaperiodate) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.