Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 0.100 N (N/10), 0.0200 M (M/50), Ricca Chemical
SDP

Catalog No. 64001SC Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Safety-coated Glass Bottle

Chemical Identifiers

CAS 7722-64-7, 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

CAS Min % 98.0, 0.31
CAS Max % 100.0, 0.32
Grade ACS/ASTM E 200/AOAC/EPA
Concentration or Composition (by Analyte or Components) 0.100N, 0.0200M
Packaging Safety-coated Glass Bottle
Quantity 4 L
Chemical Name or Material Potassium Permanganate, 0.100N, USP Volumetric Solution
ShelfLife The 10L (640025) and 20L (64005) sizes have six-month expiration (others, 18 months) due to package type.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.