Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Permanganate, 0.100 N (N/10), 0.0200 M (M/50), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Safety-coated Glass Bottle
Specifications
Specifications
CAS Min % | 98.0, 0.31 |
CAS Max % | 100.0, 0.32 |
Grade | ACS/ASTM E 200/AOAC/EPA |
Concentration or Composition (by Analyte or Components) | 0.100N, 0.0200M |
Packaging | Cubitainer |
Quantity | 10 L |
Chemical Name or Material | Potassium Permanganate, 0.100N, USP Volumetric Solution |
Safety and Handling
ShelfLife | The 10L (640025) and 20L (64005) sizes have six-month expiration (others, 18 months) due to package type. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction