Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 0.500 N (N/2), Ricca Chemical
SDP

Catalog No. p-3252197 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer

Chemical Identifiers

CAS 7722-64-7, 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Purple
CAS Min % 96.0, 1.54
CAS Max % 100.0, 1.61
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.500Normal
Chemical Name or Material Potassium Permanganate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.