Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 0.588 Normal, Reagents

Catalog No. 01336406
Change view
Click to view available options
Quantity:
1 L
4 L
Catalog No. Quantity
01-336-406 1 L
01-336-405 4 L
2 options

Catalog No. 01-336-406

Supplier: Reagents Holdings Llc C72885301C

Only null left
Add to Cart
Add to Cart

Potassium Permanganate, 0.588 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7722-64-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 101°C
Color Purple
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0126
Chemical Name or Material Potassium Permanganate, 0.588 Normal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.