Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Permanganate, 4% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
| Volume (Metric) | 500 mL |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Test Method | APHA for Hexavalent Chromium |
| Type | Potassium Permanganate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction