Learn More
Potassium permanganate, 98%
Description
Potassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.
Solubility
Soluble in water.
Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong reducing agents, powdered metals, peroxides, zinc, copper, alcohols, hydrogen fluoride and acids.
Specifications
Specifications
| Melting Point | 240°C (decomposition) |
| Quantity | 10,000 g |
| UN Number | UN1490 |
| Merck Index | 14,7655 |
| Solubility Information | Soluble in water. |
| Formula Weight | 158.03 |
| Percent Purity | 98% |
| Sensitivity | Light sensitive |
| Density | 2.7 |
| Chemical Name or Material | Potassium permanganate |
Safety and Handling
| Hazard Category | H272-H302-H314-H335-H361d-H373 |
| Hazard Statement | GHS H Statement H272-H302 May intensify fire oxidizer. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: POTASSIUM PERMANGANATE |
| EINECSNumber | 231-760-3 |
| RTECSNumber | SD6475000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.