Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium permanganate, 99+%, ACS reagent
Click to view available options
Quantity:
25 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent

Specifications
Specifications
Color | Green to Black |
Physical Form | Crystalline Powder |
Quantity | 25 g |
Assay Percent Range | 99+% |
Linear Formula | KMnO4 |
UN Number | 1490 |
Fieser | 01,942; 02,348; 04,412; 05,562; 08,416; 09,388; 10,330; 11,440; 12,413; 13,258; 14,267; 15,273; 16,283 |
Merck Index | 15, 7776 |
Formula Weight | 158.04 |
Percent Purity | ≥99.0% |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement: Very toxic to aquatic life with long lasting effects. Harmful if swallowed. May intensify fire; oxidizer. |
Precautionary Statement | GHS P Statement: Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction