Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium peroxomonosulfate, extra pure, min. 4.5% active oxygen

Catalog No. p-43392
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211361000 100 g Glass bottle
AC211360010 1 kg Glass bottle
AC211360025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC211361000 Supplier Thermo Scientific Chemicals Supplier No. 211361000
Only null left
Add to Cart

CAS: 70693-62-8 | H3K5O18S4 | 614.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70693-62-8 , 10058-23-8 , 7778-80-5 , 7646-93-7
Molecular Formula H3K5O18S4
Molecular Weight (g/mol) 614.78
MDL Number MFCD00040551
InChI Key HJKYXKSLRZKNSI-UHFFFAOYSA-I
Synonym Oxone, Potassium Monopersulfate

Specifications

Color White
Physical Form Granular Crystalline Powder
Quantity 100 g
Linear Formula KHSO4·K2SO4
Fieser 01,952; 12,413; 16,285
Solubility Information Solubility in water: 298g/L (20°C).
Formula Weight 614.78
Percent Purity ≥4.5% (active oxygen)
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Potassium peroxomonosulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 274-778-7
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.