Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Phosphate Buffer, pH 2.7, Reagents
Click to view available options
:
Quantity: 4 L
Specifications
Specifications
Chemical Name or Material | Potassium Phosphate Buffer, pH 2.7 |
Boiling Point | 100°C |
Color | Colorless |
CAS | 7732-18-5, 7778-77-0 |
Physical Form | Liquid |
Quantity | 4 L |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1.0005 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction