Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate, Dibasic, Anhydrous, FCC, 98%, Spectrum™ Chemical
SDP

Catalog No. 18605692 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail

Catalog No. 18-605-692

Supplier: Spectrum Chemical Manufacturing Corporation PO20545KGBL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

PO205, 7758-11-4, K2HPO4

Spectrum™ Chemical Potassium Phosphate, Dibasic, Anhydrous, FCC is used as an acidity regulator, antioxidant, sequestrant and stabilizer in food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
IUPAC Name dipotassium hydrogen phosphate
SMILES [K+].[K+].OP([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 45 kg
Percent Purity 98%
Grade FCC
Loss on Drying 0.02
Packaging Fiber Drum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.