Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Monobasic (Crystalline/Certified ACS), Fisher Chemical PROMO

Catalog No. P285500FB25 Shop All Fisherbrand Products
Click to view available options
:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08 g/mol
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253°C
Color White
pH 4.5
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99%
Formula Weight 136.09 amu
Percent Purity ≥99%
Grade Certified
Identification Pass Test
Loss on Drying 0.2% Max
Packaging Poly Bottle
Decomposition Information >250°C
Chemical Name or Material Potassium Phosphate Monobasic
Show More Show Less
missing translation for 'healthHazard2' CAUTION!
missing translation for 'healthHazard3'

Emergency Overview: May cause eye, skin, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1

Flammability:0

Instability:1

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.