Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phthalate ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK6704125 Shop All Avantor Products
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-6704-125 125 g Glass
MK-6704-500 500 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-6704-125 Supplier Avantor Supplier No. 670402
Add to Cart
Add to Cart

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Glass
Quantity 125 g
Formula Weight 204.22
Grade ACS
Physical Form Crystalline
Chemical Name or Material Potassium Phthalate, Primary Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.