Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium polysulfide, pure

Catalog No. AC389590010
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

CAS: 37199-66-9 | (S)nK2 | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37199-66-9
Molecular Formula (S)nK2
Molecular Weight (g/mol) NaN
MDL Number MFCD00148074
InChI Key ZOCLAPYLSUCOGI-UHFFFAOYSA-N
Synonym potassium sulfane, potassium polysulfide, caswell no. 701, potassium sulfide k2 sx, epa pesticide chemical code 076703
PubChem CID 5362529
SMILES [K+].[K+].*-[S--]-*

Specifications

Melting Point 200.0°C to 250.0°C
Quantity 1 kg
Linear Formula (K2S)n
Solubility Information Solubility in water: 500g/l (20°C)
IUPAC Name potassium sulfane
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Potassium polysulfide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 253-390-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.