Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Pyrogallate, Orsat, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Poly Bottle
Specifications
Specifications
Color | Brown |
CAS Min % | 51.0, 38.96, 7.68 |
CAS Max % | 53.0, 40.55, 8.0 |
Grade | Laboratory |
Packaging | Amber Poly Bottle |
pH | >12 |
Quantity | 500 mL |
Physical Form | Liquid |
Chemical Name or Material | Potassium Pyrogallate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction