Potassium silicate, anhydrous, Alfa Aesar™

Manufacturer: Alfa Aesar 4449322

Quantity 100g

 View more versions of this product

Catalog No. AA4449322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1312-76-1
    Physical Form Powder
    Molecular Formula SiO2:K2O 2.5:1 wt%
    MDL Number MFCD00145152
    Chemical Name or Material Potassium silicate

    Specifications

    Physical Form Powder
    Quantity 100g
    Color White
    EINECS Number 215-199-1
    Odor Odorless
    Molecular Formula SiO2:K2O 2.5:1 wt%
    MDL Number MFCD00145152
    CAS 1312-76-1
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Potassium silicate

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit