Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate Tetrahydrate, ACS Reagent (Rochelle Salt), Reagents

Catalog No. 01337791
Click to view available options
Quantity:
100 mL
500 g
3 kg
12 kg

Potassium Sodium Tartrate Tetrahydrate, ACS Reagent (Rochelle Salt), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.22
InChI Key VZOPRCCTKLAGPN-ZFJVMAEJSA-L
Synonym Rochelle Salt, Sodium potassium tartrate tetrahydrate
IUPAC Name potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
SMILES O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Boiling Point 220°C
Color Colorless/White
Physical Form Solid
Quantity 12 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 1.79
Chemical Name or Material Potassium Sodium Tartrate Tetrahydrate, ACS Reagent (Rochelle Salt)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.