Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate, Tetrahydrate, Powder, USP, 99-102%, Spectrum™ Chemical
SDP

Catalog No. 18605704 Shop All Spectrum Chemical Manufacturing Corporation Products
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail

PO210, 6381-59-5, C4H4KNaO6·4H2O

Spectrum™ Chemical Potassium Sodium Tartrate, Tetrahydrate, Powder, USP is sometimes referred to as Rochelle or Seignette's salt and used as a laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.22
InChI Key VZOPRCCTKLAGPN-ZFJVMAEJSA-L
IUPAC Name potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
SMILES O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 99 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.