Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium sorbate, 99%

Catalog No. p-4510444
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum

CAS: 24634-61-5 | C6H7KO2 | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24634-61-5
Molecular Formula C6H7KO2
Molecular Weight (g/mol) 150.22
InChI Key CHHHXKFHOYLYRE-STWYSWDKSA-M
Synonym potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
PubChem CID 23676745
ChEBI CHEBI:77868
IUPAC Name potassium;(2E,4E)-hexa-2,4-dienoate
SMILES CC=CC=CC(=O)[O-].[K+]

Specifications

Acidity Passes test
Melting Point 270.0°C
Color White to Cream
Loss on Drying 1% (105°C, 3 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3CH=CHCH=CHCOOK
Quantity 100 g
Merck Index 15, 8847
Solubility Information Solubility in water: 58.2 % (20°C). Other solubilities: soluble in alcohols: 6.5% (20°C)
Formula Weight 150.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Potassium sorbate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water
EINECSNumber 246-376-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.