Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium sorbate, 99%

Catalog No. AAA128440B
Click to view available options
Quantity:
250 g
1000 g
5000 g

CAS: 24634-61-5 | C6H7KO2 | 150.218 g/mol

Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.

Solubility
Soluble in water, alcohol, propylene glycol. Slightly soluble in acetoneand chloroform, corn oil and ether. Insoluble in benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24634-61-5
Molecular Formula C6H7KO2
Molecular Weight (g/mol) 150.218
MDL Number MFCD00016546
InChI Key CHHHXKFHOYLYRE-STWYSWDKSA-M
Synonym potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
PubChem CID 23676745
ChEBI CHEBI:77868
IUPAC Name potassium;(2E,4E)-hexa-2,4-dienoate
SMILES CC=CC=CC(=O)[O-].[K+]

Specifications

Melting Point >300°C
Density 1.361
Assay Percent Range 99%
Linear Formula CH3CH=CH-CH=CHCO2K
Quantity 1000 g
Merck Index 14,7671
Solubility Information Soluble in water,alcohol,propylene glycol. Slightly soluble in acetoneand chloroform,corn oil and ether. Insoluble in benzene.
Formula Weight 150.22
Percent Purity 99%
Chemical Name or Material Potassium Sorbate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 246-376-1
RTECSNumber WG2170000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.