Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sorbate, Powder, FCC, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18605717
Click to view available options
Quantity:
125 g
500 g
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

P1408, 590-00-1, C6H7KO2

Spectrum™ Chemical Potassium Sorbate, Powder, FCC is used as an antimicrobial preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 590-00-1
Molecular Formula C6H7KO2
Molecular Weight (g/mol) 150.22
MDL Number MFCD00016546
InChI Key CHHHXKFHOYLYRE-STWYSWDKSA-M
IUPAC Name potassium (2E,4E)-hexa-2,4-dienoate
SMILES [K+].C\C=C\C=C\C([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 98 to 101%
Grade FCC
Loss on Drying 0.01
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.