Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Standard, 1000μg/mL in water with dilute nitric acid, Ultra Scientific

Catalog No. usicc106
Encompass
Click to view available options
Quantity:
125 mL

Chemical Identifiers

CAS 7440-09-7, 7697-37-2, 7732-18-5
Molecular Formula K
Molecular Weight (g/mol) 39.10
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
PubChem CID 5462222
ChEBI CHEBI:26216
IUPAC Name potassium
SMILES [K]

Specifications

CAS Max % 99.7, 0.2, 0.1
Analyte or Component Names Water,Nitric Acid,Potassium
Element Nitric Acid, K
Solvent or Matrix Water/Dilute Nitric Acid
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Potassium Standard
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.