Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium sulfate, 99.99% (metals basis)

Catalog No. p-7049867
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate, and glass. In analytical chemistry, it is used in the determination of nitrogen by Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is also used in the separation of protein through electrophoresis. It also serves as a laboratory reagent..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689°C
Melting Point 1,069°C
Physical Form Powder
Quantity 5 g
Merck Index 14,7674
Solubility Information Soluble in water and glycerol. Insoluble in alcohol,acetate and carbon disulfide.
IUPAC Name dipotassium sulfate
Formula Weight 174.27
Percent Purity 99.99%
Odor Odorless
Density 2.662 g/mL
Assay (metals basis)
Chemical Name or Material Potassium sulfate
Show More Show Less
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-915-5
RTECSNumber TT5900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.