Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium sulfate, 99+%, for analysis, anhydrous, granular

Catalog No. AC205940025
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle

Catalog No. AC205940025

Supplier: Thermo Scientific Chemicals 205940025

Only null left
Add to Cart

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689.0°C
Melting Point 1067.0°C
Color White
pH 5.5 to 8.5 (5% soln. at 25°C)
Physical Form Granular Powder
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula K2SO4
Merck Index 15, 7793
Solubility Information Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol
IUPAC Name dipotassium sulfate
Formula Weight 174.25
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 2.66
Chemical Name or Material Potassium sulfate, p.a.
Show More Show Less
EINECSNumber 231-915-5
RTECSNumber TT5900000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.