Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulfate, For Analysis, EMSURE™, MilliporeSigma™,

Catalog No. M1051531000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Carton
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1051531000 1 kg Plastic Bottle
M1051530500 500 g Plastic Bottle
M1051535000 5 kg Plastic Bottle
M1051539025 25 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1051531000 Supplier MilliporeSigma Supplier No. 1.05153.1000
Only null left
Add to Cart
Add to Cart

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689°C (1013hPa)
Melting Point 1069°C
Color Colorless
pH 5.5 to 7.5
Physical Form Solid
Quantity 1 kg
Solubility Information 111g/L
Formula Weight 174.26
Percent Purity >99.0%
Grade ACS
Packaging Plastic Bottle
Density 2.66 g/3 (20 °C)
Show More Show Less
EINECSNumber 231-915-5
RTECSNumber TT5900000
Recommended Storage +5°C to +30°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.