Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulfate, Low Nitrogen, Reagents

Catalog No. 01337088
Change view
Click to view available options
Quantity:
500 g
3 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-088 12 kg
01-337-087 3 kg
01-337-086 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-088 Supplier Reagents Holdings Llc Supplier No. C229100012KG
Only null left
Add to Cart
Add to Cart

Potassium Sulfate, Low Nitrogen, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym Dipotassium Sulfate
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689°C
Melting Point 1069°C
Color Colorless/White
Physical Form Solid
Quantity 12 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 2.66
Chemical Name or Material Potassium Sulfate, Low Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.