Learn More
Potassium tellurite monohydrate, 97%
Description
Potassium tellurite, in association with agar is used as a selective medium for growth of clauberg (bacteria). It is used for the selective isolation of staphylococci and corynebacteria, which is used to reduce TeO3 2- to tellurium element.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 465°C (decomposition) |
Color | White |
Physical Form | Crystals |
Quantity | 25 g |
Assay Percent Range | 97% |
Solubility Information | Soluble in water. |
Formula Weight | 253.80 Anhydrous |
Appearance | White crystals |
Chemical Name or Material | Potassium tellurite monohydrate |
Safety and Handling
Hazard Category | H301 |
Precautionary Statement | P264b-P270-P301+P310-P330-P501c |
DOTInformation | DOT Class: 6.1; Packaging Group: III |
EINECSNumber | 232-213-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.