Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF, AcroSeal™

Catalog No. p-4440155
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 865-47-4 | C4H9KO | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium tert-butoxide
CAS 865-47-4, 96-47-9
Quantity 100 mL
Molecular Formula C4H9KO
Linear Formula (CH3)3COK
Flash Point -11°C
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
Solubility Information Solubility in water: reacts
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES CC(C)(C)[O-].[K+]
IUPAC Name potassium;2-methylpropan-2-olate
Molecular Weight (g/mol) 112.21
PubChem CID 23665647
Formula Weight 112.21
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.86
Density 0.8600g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.