Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tert-butoxide, pure, 20 wt.% solution in THF, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
Packaging:
Glass bottle
865-47-4,109-99-9
C4H9KO
112.213
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
23665647
potassium;2-methylpropan-2-olate
CC(C)(C)[O-].[K+]
865-47-4,109-99-9
C4H9KO
112.213
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
23665647
potassium;2-methylpropan-2-olate
CC(C)(C)[O-].[K+]
Packaging Glass bottle
Color Amber to Colorless
Specific Gravity 0.929
Flash Point −21°C
Density 0.9290g/mL
Quantity 250mL
Linear Formula (CH3)3COK
Fieser 01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
Solubility Information Solubility in water: reacts with water.
Formula Weight 112.21
Physical Form Solution
Grade Pure
Chemical Name or Material Potassium tert-butoxide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapour. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger