Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetraborate tetrahydrate, 99.5%, extra pure

Catalog No. p-4510344
Click to view available options
Quantity:
250 g
1 kg
5 kg

CAS: 12045-78-2 | B4K2O7·4H2O | 305.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12045-78-2
Molecular Formula B4K2O7·4H2O
Molecular Weight (g/mol) 305.5
Synonym Potassium biborate tetrahydrate

Specifications

Melting Point 850.0°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.5%
Linear Formula K2B4O7·4H2O
Merck Index 14,768
Solubility Information Solubility in water: soluble.
Formula Weight 305.5
Percent Purity 99.5%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Potassium tetraborate tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.