Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetraborate tetrahydrate, 99%

Catalog No. AA39435A3
Click to view available options
Quantity:
500 g
2 kg

CAS: 12045-78-2 | K2B4O7+4H2O

It is an excellent solvent for metallic oxides at high temperatures. It is a good buffering agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12045-78-2
Molecular Formula K2B4O7+4H2O
MDL Number MFCD00149924
Synonym Potassium borate (tetra)

Specifications

Melting Point (decomposition)
Color White
Physical Form Granules
Quantity 2 kg
Assay Percent Range 99%
Solubility Information Soluble in water
Formula Weight 305.51 (233.44 Anhydrous)
Odor Odorless
Appearance White crystals
Density 1.74 (anhy)
Chemical Name or Material Potassium tetraborate tetrahydrate
Show More Show Less
Hazard Category H361d
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 215-575-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.