Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrabromoaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 33% min

Catalog No. AA1266403
Click to view available options
Quantity:
1 g
5 g

CAS: 13005-38-4 | KAuBr4·2H2O

It is used as a Laboratory chemical and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13005-38-4
Molecular Formula KAuBr4·2H2O
MDL Number MFCD00049650
Synonym Gold potassium bromide

Specifications

Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water and alcohol
Formula Weight 591.72 (555.69 Anhydrous)
Odor Weak
Appearance Red-brown crystals
Chemical Name or Material Potassium tetrabromoaurate(III) dihydrate, Au 33% min.
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-268-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.