Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrachloroaurate(III), 98%

Catalog No. ac316120010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 13682-61-6 | AuCl4K | 377.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13682-61-6
Molecular Formula AuCl4K
Molecular Weight (g/mol) 377.88
MDL Number MFCD00043088
InChI Key JMONCQYBAMKVQQ-UHFFFAOYSA-J
Synonym potassium tetrachloroaurate iii, potassium tetrachloroaurate, k aucl4, kaucl4 * 2h2o, potassium tetrachloridoaurate, potassium ion tetrachloroaurate, potassium tetrachloroaurate 1-, potassium tetrachloridoaurate 1-, potassium tetrachloridoaurate iii, potassium gold iii chloride
PubChem CID 11337898
ChEBI CHEBI:51564
IUPAC Name potassium;tetrachlorogold(1-)
SMILES Cl[Au-](Cl)(Cl)Cl.[K+]

Specifications

Melting Point >300.0°C
Color Yellow to Orange
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 97.5% min
Linear Formula KAuCl4
Merck Index 15, 7800
Solubility Information Solubility in water: soluble
Formula Weight 377.88
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Potassium tetrachloroaurate(III)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-190-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.