Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrachloropalladate(II), min 32.0% Pd

Catalog No. p-43548
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle

CAS: 10025-98-6 | Cl4K2Pd | 326.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-98-6
Molecular Formula Cl4K2Pd
Molecular Weight (g/mol) 326.41
MDL Number MFCD00011373
InChI Key LGCKLDWLSVFMGL-UHFFFAOYSA-J
Synonym potassium tetrachloropalladate ii, potassium chloropalladite, potassium tetrachloropalladate, ccris 6625, potassium chlorophalladite, potassium palladium chloride, potassium palladous chloride, dipotassium tetrachloropalladate ii, dipotassium tetrachloropalladate 2-, tetrachloropalladate 2-dipotassium
PubChem CID 61438
IUPAC Name dipotassium;tetrachloropalladium(2-)
SMILES Cl[Pd-2](Cl)(Cl)Cl.[K+].[K+]

Specifications

Melting Point 105.0°C
Color Brown to Brown-Green
Physical Form Crystalline Powder and/or Crystals
Quantity 2 g
Linear Formula K2PdCl4
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 326.41
Percent Purity ≥32% (Pd)
Packaging Glass bottle
Specific Gravity 2.67
Density 2.6700g/mL
Chemical Name or Material Potassium tetrachloropalladate(II), min 32.0% Pd
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 233-049-3
RTECSNumber TT4100000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.