Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium tetrachloroplatinate(II), Premion™, 99.99% (metals basis), Pt 46.4% min

Manufacturer:  Alfa Aesar™ 4394603

Quantity 1g

 View more versions of this product

Catalog No. AA4394603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10025-99-7
K2PtCl4
MFCD00011378
14,7682
Premion™
Cl4K2Pt
415.11
UN3288
Potassium tetrachloroplatinate (II)

Specifications

10025-99-7
Cl4K2Pt
415.11
UN3288
Pt 46.4% min
Odorless
1g
99.99%, (metals basis)
Crystalline
K2PtCl4
MFCD00011378
14,7682
3.38g/mL
Red-brown
Potassium tetrachloroplatinate (II)
Premion™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H334-H318-H290-H315-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P301+P310-P305+P351+P338-P405-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : TP1850000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit