Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetracyanonickelate(II) hydrate

Catalog No. p-7050771
Click to view available options
Quantity:
10 g
50 g

CAS: 339527-86-5 | C4K2N4Ni | 240.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 339527-86-5
Molecular Formula C4K2N4Ni
Molecular Weight (g/mol) 240.96
MDL Number MFCD00049669
InChI Key QOOHKAMDWOKKBR-UHFFFAOYSA-N
Synonym potassium tetracyanonickelate ii hydrate, c4n4ni.2k.h2o, dipotassium tetracyanonickelate 2-hydrate
IUPAC Name nickel(2+) dipotassium tetrakis(iminomethanide) hydrate
SMILES [K+].[K+].N#C[Ni--](C#N)(C#N)C#N

Specifications

Melting Point 100°C (in -H2O)
Physical Form Crystalline
Quantity 10 g
UN Number UN1588
Merck Index 14,7684
Solubility Information Soluble in water
Formula Weight 240.99 (Anhydrous)
Density 1.875g/mL
Chemical Name or Material Potassium tetracyanonickelate(II) hydrate
Hazard Category H300+H310+H330-H317-H334-H350i
Hazard Statement GHS H Statement
H301-H334-H350-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P262-P264b-P270-P271-P272-P280g-P281-P284-P285-P301+P310-P302+P350-P304+P340-P308+P313-P310-P330-P333+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 238-082-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.