Learn More
Potassium tetrafluoroborate, 98%
Description
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.
Solubility
Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol.
Notes
Moisture sensitive. Incompatible withstrong oxidizing agents.

Specifications
Specifications
Boiling Point | (decomposition) |
Melting Point | 529°C |
Quantity | 1000 g |
Linear Formula | KBF4 |
Merck Index | 14,7686 |
Solubility Information | Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol. |
Formula Weight | 125.91 |
Percent Purity | 98% |
Refractive Index | 1.3245 |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
EINECSNumber | 237-928-2 |
RTECSNumber | ED2800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.