Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrafluoroborate, 99%, extra pure

Catalog No. p-43577
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 14075-53-7 | BF4K | 125.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14075-53-7
Molecular Formula BF4K
Molecular Weight (g/mol) 125.90
MDL Number MFCD00011395
InChI Key AKEBROIVCDHVSD-UHFFFAOYSA-N
Synonym potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride
PubChem CID 518872
IUPAC Name potassium tetrafluoroboranuide
SMILES [K+].F[B-](F)(F)F

Specifications

Melting Point 530.0°C
Color White to Gray
Physical Form Micro-crystalline Powder
Quantity 25 g
Assay Percent Range 98.5% min
Linear Formula KBF4
Merck Index 15,7804
Solubility Information Solubility in water: 4.4g/L (20°C). Other solubilities: slightly soluble in boiling alcohol
Formula Weight 125.91
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Density 2.5
Chemical Name or Material Potassium tetrafluoroborate, c.p.
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-928-2
RTECSNumber ED2800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.