Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Tetrakis(4-chlorophenyl)borate 98.0+%, TCI America™
SDP

Catalog No. A51325G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 14680-77-4
Molecular Formula C24H16BCl4K
Molecular Weight (g/mol) 496.10
MDL Number MFCD00043105
InChI Key VHPYLDPFVZBNQT-UHFFFAOYSA-N
Synonym potassium tetrakis 4-chlorophenyl borate, tetrakis 4-chlorophenyl boron potassium, k-tcpb, potassium tetrakis 4-chlorophenyl boranuide, potassium tetrakis 4-chlorophenyl borate, selectophore tm
PubChem CID 23679691
IUPAC Name boron(3+) potassium tetrakis(3-chlorobenzen-1-ide)
SMILES [B+3].[K+].ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1

Specifications

Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 496.10
Percent Purity ≥98.0% (HPLC,W)
Chemical Name or Material Potassium Tetrakis(4-chlorophenyl)borate [Anion for the neutral carrier type ion electrode]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.