Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrakis(pentafluorophenyl)borate, 97%

Catalog No. p-7032985
Click to view available options
Quantity:
1 g
5 g

CAS: 89171-23-3 | C24BF20K | 718.14 g/mol

Potassium tetrakis(pentafluorophenyl)borate is used as a biochemical reagent, a catalyst and a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrakis(pentafluorophenyl)borate is used as a biochemical reagent, a catalyst and a pharmaceutical intermediate.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89171-23-3
Molecular Formula C24BF20K
Molecular Weight (g/mol) 718.14
MDL Number MFCD06797410
InChI Key GYBHRIJOPWTIKA-UHFFFAOYSA-N
Synonym potassium tetrakis pentafluorophenyl borate, potassium tetrakis pentafluorophe-nyl borate, tetrakis-pentafluorophenyl-potassium borate, potassium tetrakis 2,3,4,5,6-pentafluorophenyl boranuide
PubChem CID 23693577
IUPAC Name potassium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
SMILES [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.[K+]

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 718.13
Percent Purity 97%
Chemical Name or Material Potassium tetrakis(pentafluorophenyl)borate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 438-590-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.