Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium thiocyanate, ACS, 98.5% min

Catalog No. AA1431836
Click to view available options
Quantity:
100 g
500 g

CAS: 333-20-0 | CKNS | 97.176 g/mol

Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the convertion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. Also used in moisturizers and oral care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Boiling Point 500°C (decomposition)
Melting Point 172°C to 175°C
Color White
Physical Form Crystals
Quantity 500 g
Assay Percent Range 98.5% min.
Beilstein 3594799
Solubility Information Soluble in water,acetone,and alcohol. When dissolved in its own weight of water,the temperature drops ^=30° Soluble in water,acetone and alcohol.
Formula Weight 97.18
Grade ACS
Odor Odorless
Sensitivity Light and moisture sensitive
Appearance White crystalline
Density 1.89
Chemical Name or Material Potassium thiocyanate, For ACS analysis
Show More Show Less
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 206-370-1
RTECSNumber XL1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.