Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Thiocyanate, Purified, Reagents

Catalog No. 01337084
Click to view available options
Quantity:
125 g
500 g
2.5 kg

Potassium Thiocyanate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.18
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym Potassium Rhodanide, Potassium Rhodanate
IUPAC Name potassium cyanosulfanide
SMILES [K+].[S-]C#N

Specifications

Boiling Point 500°C
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.89
Chemical Name or Material Potassium Thiocyanate, Purified
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.