Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Thiocyanate, Purified, Reagents

Catalog No. 01337085
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Catalog No. Quantity
01-337-085 125 g
01-337-084 2.5 kg
01-337-083 500 g
3 options

Catalog No. 01-337-085

Supplier: Reagents Holdings Llc C2293000125D2

Only null left
Add to Cart
Add to Cart

Potassium Thiocyanate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.18
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym Potassium Rhodanide, Potassium Rhodanate
IUPAC Name potassium cyanosulfanide
SMILES [K+].[S-]C#N

Specifications

Boiling Point 500°C
Color Colorless/White
Physical Form Solid
Quantity 125 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 1.89
Chemical Name or Material Potassium Thiocyanate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.