Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium thiophene-3-trifluoroborate, 97%, Thermo Scientific™

Catalog No. p-7052689
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192863-37-9
Molecular Formula C4H3BF3KS
Molecular Weight (g/mol) 190.032
MDL Number MFCD03701626
InChI Key ULOOPHJJEPUNNH-UHFFFAOYSA-N
Synonym potassium 3-thiophenetrifluoroborate, potassium thiophene-3-trifluoroborate, potassium trifluoro thiophen-3-yl boranuide, potassium trifluoro thiophen-3-yl borate, pubchem11540, amtb173, potassiurn 3-thiophenetri tluoroborate, potassiotrifluoro 3-thienyl boron v, potassium trifluoro 3-thiophenyl boranuide, potassium trifluoro thiophen-3-yl borate 1-
PubChem CID 23677379
IUPAC Name potassium;trifluoro(thiophen-3-yl)boranuide
SMILES [B-](C1=CSC=C1)(F)(F)F.[K+]

Specifications

Color White
Physical Form Powder
Quantity 250 mg
Formula Weight 190.04
Percent Purity 97%
Chemical Name or Material Potassium thiophene-3-trifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber ED3367500
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.