Learn More
Potassium tricyanomethanide
Description
Potassium tricyanomethanide is a precursor used for the preparation of dicyano ketene aminals. It reacts with amines to prepare alkyl ammonium tricyanomethanide. Further, it is involved in the preparation of tricyanomethanide-bridged binuclear organometallic complexes such as Cp(dppe)FeC(CN)3 and Cp(PPh3)2RuC(CN)3. It is used as starting material utilized in the preparation of 5-Amino-3-bromo-1-(tert-butyl)-1H-pyrazole-4-carbonitrile, which undergoes sandmeyer reaction to get the corresponding diaminopyrazole, 5-amino-3-aryl-1-(tert-butyl)-1H-pyrazole-4-carboxamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Linear Formula | KC(CN)3 |
| Quantity | 1 g |
| UN Number | UN3439 |
| Solubility Information | Soluble in water. |
| Formula Weight | 129.16 |
| Physical Form | Crystalline |
| Chemical Name or Material | Potassium tricyanomethanide |
Safety and Handling
| Hazard Category | H301+H311-H330 |
| Hazard Statement | GHS H Statement H300-H310-H330 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.